Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198839.1 | NP_001185768.1 | 5 | 1419 | ||
NM_009954.3 | NP_034084.2 | 5 | 1482 | |||
GenBank mRNA | AK079554.1 | - | 4 | 1444 | ||
AK090315.1 | - | 5 | 1492 | |||
AK145863.1 | - | 5 | 1483 | |||
AK159168.1 | - | 5 | 1386 | |||
AK159297.1 | - | 5 | 1387 | |||
AK166248.1 | - | 5 | 1357 | |||
AK167352.1 | - | 5 | 1420 | |||
AK167504.1 | - | 5 | 1420 | |||
AK167516.1 | - | 5 | 1484 | |||
AK167717.1 | - | 5 | 1483 | |||
BC057578.1 | - | 5 | 1442 | 6401186 | ||
U28151.1 | - | 5 | 1393 | |||
U48853.1 | - | 5 | 1377 |