Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009959.1 | NP_034089.1 | 1 | 779 | ||
GenBank mRNA | AB008181.1 | - | 1 | 672 | ||
AY013763.1 | - | 1 | 779 |