Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009965.2 | NP_034095.1 | 6 | 670 | ||
GenBank mRNA | AJ239052.1 | - | 6 | 670 | ||
AK084145.1 | - | 3 | 1247 | |||
BC107342.2 | - | 6 | 670 | 40055280 | ||
J00378.1 | - | 3 | 443 | |||
V00728.1 | - | 4 | 562 |