Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009969.4 | NP_034099.2 | 4 | 900 | ||
GenBank mRNA | AK053196.1 | - | 4 | 900 | ||
X02333.1 | - | 4 | 785 | |||
X03019.1 | - | 4 | 643 | |||
X03221.1 | - | 4 | 642 | |||
X05906.1 | - | 4 | 542 |