Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009982.4 | NP_034112.3 | 4 | 646 | ||
GenBank mRNA | AK002454.1 | - | 4 | 605 | ||
AK049406.1 | - | 4 | 605 | |||
AK146858.1 | - | 4 | 654 | |||
AK150761.1 | - | 4 | 606 | |||
AK151388.1 | - | 4 | 605 | |||
AK151711.1 | - | 4 | 606 | |||
AK151861.1 | - | 4 | 492 | |||
AK153168.1 | - | 4 | 606 | |||
AK167882.1 | - | 4 | 605 | |||
BC067063.1 | - | 4 | 579 | 30541061 | ||
U74683.3 | - | 4 | 603 | |||
U89269.1 | - | 4 | 604 |