Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009993.3 | NP_034123.1 | 7 | 1769 | ||
GenBank mRNA | BC018298.1 | - | 7 | 1749 | 4196676 | |
BC054827.1 | - | 7 | 1763 | 5054733 | ||
K02589.1 | - | 6 | 1770 | |||
X00479.1 | - | 7 | 1770 | |||
X04283.1 | - | 6 | 1732 |