Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp24a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009996.3 | NP_034126.1 | 12 | 2420 | ||
GenBank mRNA | AK016668.1 | - | 1 | 164 | ||
AK143984.1 | - | 12 | 2428 | |||
AK159109.1 | - | 12 | 2428 | |||
AK159481.1 | - | 12 | 2427 | |||
AK159527.1 | - | 12 | 2427 | |||
AK159852.1 | - | 12 | 2428 | |||
AK159954.1 | - | 12 | 2428 | |||
AK159976.1 | - | 12 | 2428 | |||
D89669.1 | - | 12 | 2420 |