Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010004.2 | NP_034134.2 | 2 | 316 | ||
GenBank mRNA | AF047727.1 | - | 2 | 230 | ||
BC139471.1 | - | 2 | 316 | 8862493 | ||
BC139472.1 | - | 2 | 316 | 8862494 |
Gene Symbol: Cyp2c69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104525.1 | NP_001097995.1 | 2 | 286 | ||
GenBank mRNA | BC145630.1 | - | 2 | 316 |