Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF090317.1 | - | 1928 | |||
BC010973.1 | - | 1952 | 4161038 | |||
BC010974.1 | - | 1945 | 4162964 | |||
BC049969.1 | - | 1927 | 4158774 |