Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276481.1 | NP_001263410.1 | 2 | 698 | ||
NM_001276482.1 | NP_001263411.1 | 3 | 611 | |||
NM_001276485.1 | NP_001263414.1 | 2 | 542 | |||
NM_001276486.1 | NP_001263415.1 | 3 | 599 | |||
NM_001276492.1 | NP_001263421.1 | 4 | 746 | |||
NM_001276493.1 | NP_001263422.1 | 3 | 703 | |||
NM_001276494.1 | NP_001263423.1 | 3 | 594 | |||
NM_010017.4 | NP_034147.1 | 3 | 755 | |||
GenBank mRNA | AK036174.1 | - | 3 | 566 | ||
AK038702.1 | - | 3 | 677 | |||
AK053886.1 | - | 2 | 503 | |||
AK145048.1 | - | 3 | 560 | |||
BC007150.1 | - | 3 | 568 | 3496914 | ||
CA327718.1 | - | 2 | 655 | |||
CD578569.1 | - | 3 | 570 | |||
X86073.1 | - | 1 | 296 |