Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010023.4 | NP_034153.2 | 7 | 899 | ||
GenBank mRNA | AK004838.1 | - | 7 | 900 | ||
AK029481.1 | - | 7 | 900 | |||
AK077386.1 | - | 7 | 890 |