Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109750.1 | NP_001103220.1 | 4, 5 | 809 | ||
NM_010048.3 | NP_034178.2 | 4, 5 | 818 | |||
GenBank mRNA | AK042786.1 | - | 4, 5 | 740 | ||
AK088131.1 | - | 4, 5 | 739 | |||
AK129070.1 | - | 4, 5 | 750 | |||
AK133880.1 | - | 1, 2 | 107 | |||
AK141594.1 | - | 4, 5 | 784 | |||
AK145495.1 | - | 4, 5 | 735 | |||
AK153026.1 | - | 4, 5 | 606 | |||
AK161022.1 | - | 1569 | ||||
AK161359.1 | - | 4, 5 | 534 | |||
AK163872.1 | - | 4, 5 | 771 | |||
AK163903.1 | - | 4, 5 | 746 | |||
AK201465.1 | - | 2, 3 | 267 | |||
BC060636.1 | - | 4, 5 | 759 | 6849298 | ||
BC062978.1 | - | 4, 5 | 713 | 30356000 | ||
D78641.1 | - | 4, 5 | 743 | |||
X95480.1 | - | 4, 5 | 684 |