Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010050.2 | NP_034180.1 | 2 | 903 | ||
GenBank mRNA | AF093137.1 | - | 2 | 761 | ||
AF096875.1 | - | 2 | 903 | |||
AF177196.1 | - | 2 | 773 | |||
AK032038.1 | - | 2 | 613 | |||
AK045607.1 | - | 2 | 657 | |||
BC125383.1 | - | 2 | 798 | 40129898 | ||
BC125385.1 | - | 2 | 798 | 40129900 |