Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010060.3 | NP_034190.3 | 3 | 690 | ||
GenBank mRNA | AF183144.1 | - | 3 | 690 |