Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trdmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010067.4 | NP_034197.3 | 5, 6 | 431 | ||
GenBank mRNA | AF012129.1 | - | 5, 6 | 418 | ||
AF045889.1 | - | 5, 6 | 423 | |||
AK029136.1 | - | 5, 6 | 432 | |||
AK050403.1 | - | 5, 6 | 431 | |||
BC087892.1 | - | 5, 6 | 431 | 30504522 |