Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005402.3 | NP_005393.2 | 3 | 498 | ||
XM_006715762.2 | XP_006715825.1 | 3 | 290 | |||
XM_011515466.1 | XP_011513768.1 | 4 | 542 | |||
GenBank mRNA | AF493910.1 | - | 2 | 197 | ||
AK308109.1 | - | 3 | 351 | |||
AU132355.1 | - | 3 | 488 | |||
BC039858.1 | - | 3 | 444 | 5495399 | ||
DC313757.1 | - | 3 | 498 | |||
M29893.1 | - | 2 | 197 | |||
X15014.1 | - | 2 | 188 |