Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113414.1 | NP_001106885.1 | 3 | 851 | ||
NM_001113415.1 | NP_001106886.1 | 3 | 851 | |||
NM_010096.3 | NP_034226.1 | 3 | 851 | |||
GenBank mRNA | AK220542.1 | - | 3 | 851 | ||
BC067018.1 | - | 4 | 606 | 6833120 | ||
U82442.1 | - | 2 | 148 | |||
U92702.1 | - | 3 | 392 | |||
U92704.1 | - | 3 | 392 |