Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sparcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010097.4 | NP_034227.3 | 3 | 809 | ||
GenBank mRNA | AK134608.1 | - | 3 | 450 | ||
AK158725.1 | - | 3 | 316 | |||
BC003759.1 | - | 3 | 454 | 3486162 | ||
U64827.1 | - | 3 | 407 | |||
U77330.1 | - | 3 | 474 |