Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIK1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243532.1 | NP_001230461.1 | 2 | 512 | ||
NM_001243533.1 | NP_001230462.1 | 3 | 586 | |||
NM_152835.4 | NP_690048.1 | 2 | 395 | |||
XM_005245743.4 | XP_005245800.1 | 3 | 655 | |||
GenBank mRNA | AF411102.1 | - | 3 | 526 | ||
AK312872.1 | - | 2 | 395 | |||
BC028713.1 | - | 3 | 528 | 4828997 | ||
CV024336.1 | - | 1 | 239 | |||
DA221039.1 | - | 2 | 467 | |||
DB227864.1 | - | 2 | 395 | |||
DB233669.1 | - | 2 | 512 |