Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013469.2 | NP_038497.2 | 5 | 721 | ||
GenBank mRNA | AK044203.1 | - | 5 | 694 | ||
AK137158.1 | - | 5 | 665 | |||
AK140375.1 | - | 5 | 694 | |||
AK170850.1 | - | 4 | 1739 | |||
BC012875.1 | - | 5 | 641 | 3500318 | ||
CT010331.1 | - | 4 | 572 | |||
U65986.1 | - | 5 | 706 |