Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013469.2 | NP_038497.2 | 14 | 1562 | ||
GenBank mRNA | AK137158.1 | - | 13 | 1872 | ||
AK140375.1 | - | 14 | 1535 | |||
BC012875.1 | - | 14 | 1482 | 3500318 | ||
CT010331.1 | - | 13 | 1413 | |||
U65986.1 | - | 14 | 1547 |