Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XRCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005431.1 | NP_005422.1 | 3 | 251 | ||
GenBank mRNA | AF035587.1 | - | 3 | 251 | ||
AK313607.1 | - | 3 | 227 | |||
BC042137.1 | - | 3 | 226 | 5801996 | ||
CR749256.1 | - | 3 | 187 | |||
Y08837.1 | - | 3 | 247 |