Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013488.2 | NP_038516.1 | 10 | 1535 | ||
GenBank mRNA | AK153966.1 | - | 10 | 1535 | ||
AK161775.1 | - | 10 | 1459 | |||
BC039137.1 | - | 10 | 1514 | 3670597 | ||
M13816.1 | - | 9 | 1405 | |||
M36850.1 | - | 9 | 1405 | |||
X04836.1 | - | 10 | 1535 |