Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013488.2 | NP_038516.1 | 10 | 2120 | ||
GenBank mRNA | AK153966.1 | - | 10 | 2121 | ||
AK161775.1 | - | 10 | 2044 | |||
BC039137.1 | - | 10 | 2099 | 3670597 | ||
X04836.1 | - | 10 | 2121 |