Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 3 | 493 | ||
NM_001198860.1 | NP_001185789.1 | 4 | 686 | |||
NM_001198861.1 | NP_001185790.1 | 4 | 686 | |||
NM_013502.3 | NP_038530.1 | 3 | 493 | |||
GenBank mRNA | AB033122.1 | - | 3 | 458 | ||
AJ010483.1 | - | 3 | 406 | |||
AK133816.1 | - | 4 | 657 | |||
AK149906.1 | - | 4 | 664 | |||
AK160658.1 | - | 4 | 676 | |||
AK165276.1 | - | 4 | 641 | |||
AK170133.1 | - | 2 | 200 | |||
AK171650.1 | - | 2 | 326 | |||
AK215609.1 | - | 94 | ||||
AK217104.1 | - | 2 | 220 | |||
AK218288.1 | - | 2 | 205 | |||
BC013702.1 | - | 3 | 406 | 3155102 | ||
BC015071.1 | - | 3 | 462 | 4192052 | ||
BC042425.1 | - | 4 | 667 | 5005681 |