Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 9 | 1496 | ||
NM_001198860.1 | NP_001185789.1 | 10 | 1689 | |||
NM_001198861.1 | NP_001185790.1 | 10 | 1692 | |||
NM_013502.3 | NP_038530.1 | 9 | 1499 | |||
GenBank mRNA | AB033122.1 | - | 9 | 1464 | ||
AJ010483.1 | - | 9 | 1409 | |||
AK133816.1 | - | 10 | 1663 | |||
AK149906.1 | - | 7 | 3106 | |||
AK160658.1 | - | 10 | 1679 | |||
AK165276.1 | - | 10 | 1647 | |||
AK170133.1 | - | 8 | 1206 | |||
AK171650.1 | - | 8 | 1332 | |||
BC013702.1 | - | 9 | 1412 | 3155102 | ||
BC015071.1 | - | 9 | 1468 | 4192052 | ||
BC042425.1 | - | 10 | 1673 | 5005681 |