Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032054.1 | - | 1 | 657 | ||
AK045456.1 | - | 1 | 846 | |||
AY255563.1 | - | 1 | 355 | |||
BC138059.1 | - | 1 | 714 | 8861078 | ||
BC138060.1 | - | 1 | 714 | 8861079 |