Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013803.2 | NP_038831.2 | 3 | 930 | ||
GenBank mRNA | AF110178.1 | - | 3 | 931 | ||
AF110179.1 | - | 3 | 931 | |||
AF128842.1 | - | 2 | 628 |
Set Membership: |
G-protein coupled receptor |