Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013805.4 | NP_038833.2 | 1 | 1211 | ||
GenBank mRNA | AF035814.1 | - | 1 | 1203 | ||
AK077282.1 | - | 1 | 1212 | |||
BC002016.1 | - | 1 | 1195 | 3488769 | ||
BC083341.1 | - | 1 | 1201 | 6819173 |