Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015734.2 | NP_056549.2 | 66 | 6183 | ||
GenBank mRNA | AB009993.1 | - | 66 | 5923 | ||
AK148055.1 | - | 23 | 2326 | |||
AK157100.1 | - | 3 | 843 |