Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bhmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016668.3 | NP_057877.1 | 5 | 796 | ||
GenBank mRNA | AF033381.1 | - | 5 | 539 | ||
AK077108.1 | - | 5 | 603 | |||
AK145668.1 | - | 5 | 638 | |||
AK149431.1 | - | 4 | 940 | |||
AK149457.1 | - | 5 | 603 | |||
AK166754.1 | - | 5 | 605 | |||
AK166818.1 | - | 5 | 604 | |||
BC037004.1 | - | 5 | 598 | 5100429 | ||
BC093510.1 | - | 5 | 571 | 5101184 | ||
BC110307.1 | - | 5 | 999 | 5310035 | ||
BG078929.2 | - | 3 | 288 |