Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016742.4 | NP_058022.1 | 4 | 617 | ||
GenBank mRNA | AK010494.1 | - | 4 | 642 | ||
AK013255.1 | - | 4 | 618 | |||
AK145671.1 | - | 4 | 649 | |||
AK168651.1 | - | 4 | 635 | |||
BC060079.1 | - | 4 | 640 | 5714770 | ||
U43076.1 | - | 4 | 655 |