Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079812.2 | NP_001073280.1 | 2 | 305 | ||
NM_001314007.1 | NP_001300936.1 | 3 | 332 | |||
NM_005219.4 | NP_005210.3 | 3 | 332 | |||
XM_011537572.2 | XP_011535874.1 | 2 | 273 | |||
XM_011537573.2 | XP_011535875.1 | 3 | 140 | |||
GenBank mRNA | AB209482.1 | - | 3 | 273 | ||
AF051782.1 | - | 2 | 164 | |||
AK304291.1 | - | 3 | 278 | |||
AY363395.1 | - | 3 | 191 | |||
BC117257.1 | - | 2 | 182 | 40125808 | ||
BC143413.1 | - | 3 | 209 | 9051921 | ||
BC143414.1 | - | 3 | 209 | |||
BQ945734.1 | - | 3 | 266 | |||
DA593734.1 | - | 3 | 332 | |||
DQ067453.1 | - | 2 | 179 |