Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016779.2 | NP_058059.2 | 5, 6 | 305 | ||
GenBank mRNA | AK132177.1 | - | 5, 6 | 306 | ||
AK157973.1 | - | 5, 6 | 306 | |||
BC113753.1 | - | 5, 6 | 227 | 40039844 | ||
U65020.1 | - | 5, 6 | 311 |