Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 15 | 1780 | ||
NM_016878.4 | NP_058574.3 | 15 | 1604 | |||
GenBank mRNA | AF005051.1 | - | 14 | 1714 | ||
AK038646.1 | - | 1780 | ||||
AK052001.1 | - | 2171 | ||||
AK160084.1 | - | 15 | 1541 | |||
AK166276.1 | - | 15 | 1554 | |||
AK209838.1 | - | 1 | 76 | |||
AK213377.1 | - | 1 | 76 | |||
BC092232.1 | - | 15 | 1532 | 30551654 |