Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016892.3 | NP_058588.1 | 8 | 937 | ||
GenBank mRNA | AF173379.1 | - | 8 | 910 | ||
AK010264.1 | - | 5 | 546 | |||
AK050072.1 | - | 8 | 886 | |||
AK205191.1 | - | 1 | 71 | |||
BC026938.1 | - | 8 | 904 | 4024140 | ||
CF929049.1 | - | 2 | 263 |