Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017367.3 | NP_059063.2 | 7 | 2531 | ||
GenBank mRNA | AK079476.1 | - | 7 | 2472 | ||
AK152614.1 | - | 7 | 2488 | |||
BC003290.1 | - | 7 | 2064 | 3583418 | ||
BC029328.1 | - | 7 | 2064 | |||
CA451138.1 | - | 1 | 51 |