Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276684.1 | NP_001263613.1 | 1 | 1680 | ||
NM_018790.3 | NP_061260.1 | 1 | 1680 | |||
GenBank mRNA | AF162777.1 | - | 1 | 1671 | ||
AK157822.1 | - | 1 | 1681 | |||
BC023127.1 | - | 1 | 1669 | 3498057 |