Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019388.3 | NP_062261.3 | 8 | 2290 | ||
GenBank mRNA | AK079513.1 | - | 8 | 2291 | ||
AK170054.1 | - | 8 | 2291 | |||
BC013807.1 | - | 8 | 2262 | 4008635 |