Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019707.4 | NP_062681.2 | 2, 3 | 251 | ||
GenBank mRNA | AB022100.1 | - | 2, 3 | 260 | ||
AK016527.1 | - | 3, 4 | 1509 | |||
AK029195.1 | - | 2, 3 | 210 | |||
AK032409.1 | - | 2, 3 | 280 | |||
AK034995.1 | - | 2, 3 | 231 | |||
AK039438.1 | - | 2, 3 | 273 | |||
AK045201.1 | - | 2, 3 | 252 | |||
AK048724.1 | - | 2, 3 | 251 | |||
AK085939.1 | - | 2, 3 | 211 | |||
AK134649.1 | - | 2, 3 | 210 | |||
BC021628.1 | - | 2, 3 | 265 | 3987627 |