Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019707.4 | NP_062681.2 | 3 | 296 | ||
GenBank mRNA | AB022100.1 | - | 3 | 305 | ||
AK016527.1 | - | 4 | 1554 | |||
AK029195.1 | - | 3 | 255 | |||
AK032409.1 | - | 3 | 325 | |||
AK034995.1 | - | 3 | 276 | |||
AK039438.1 | - | 3 | 318 | |||
AK045201.1 | - | 3 | 297 | |||
AK048724.1 | - | 3 | 296 | |||
AK085939.1 | - | 3 | 256 | |||
AK134649.1 | - | 3 | 255 | |||
BC021628.1 | - | 3 | 310 | 3987627 |