Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005261.3 | NP_005252.1 | 4 | 793 | ||
NM_181702.2 | NP_859053.1 | 4 | 678 | |||
XM_017013315.1 | XP_016868804.1 | 4 | 819 | |||
GenBank mRNA | AK314017.1 | - | 4 | 678 | ||
BC022010.2 | - | 4 | 678 | 4823985 | ||
U10550.1 | - | 4 | 767 | |||
U13052.1 | - | 3 | 554 |