Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020260.2 | NP_064656.2 | 1 | 436 | ||
GenBank mRNA | AF151363.1 | - | 1 | 77 | ||
AK018057.1 | - | 1 | 302 | |||
AK045213.1 | - | 1 | 300 | |||
AK136348.1 | - | 1 | 656 | |||
AK138417.1 | - | 1 | 676 | |||
AK149975.1 | - | 1 | 272 | |||
AK150109.1 | - | 1 | 272 | |||
AK155729.1 | - | 1 | 256 | |||
AK170685.1 | - | 1 | 256 | |||
BB865916.1 | - | 1 | 438 | |||
BC138870.1 | - | 1 | 241 | 8861892 |