Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbil5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021294.2 | NP_067269.1 | 2 | 343 | ||
GenBank mRNA | AK006528.1 | - | 2 | 304 | ||
BC048474.1 | - | 2 | 322 | 6773240 | ||
CN833562.1 | - | 346 | ||||
S83465.1 | - | 2 | 330 |