Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021353.3 | NP_067328.1 | 16, 17 | 1970 | ||
GenBank mRNA | AF136751.1 | - | 16, 17 | 1888 | ||
BC139271.1 | - | 15, 16 | 1805 | 8862293 | ||
BC139273.1 | - | 15, 16 | 1805 | 8862295 |