Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021456.4 | NP_067431.2 | 12 | 1461 | ||
GenBank mRNA | AK136446.1 | - | 12 | 1458 | ||
AK136588.1 | - | 12 | 1458 | |||
AK149446.1 | - | 12 | 1458 | |||
AK162328.1 | - | 12 | 1436 | |||
BC021150.1 | - | 12 | 1480 | 4163576 | ||
BC026897.1 | - | 12 | 1441 | 4195247 | ||
Y12887.1 | - | 12 | 1447 |