Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCDH15 SIRNA Assays ›
Gene Symbol:
Pcdh15
Gene Name:
protocadherin 15
Gene Aliases:
BB078305, ENSMUSG00000046980, Gm9815, Ush1f, av, nmf19
Chromosome Location:
Chr.10: 73100028 - 74649737 on Build GRCm38
UniGene ID:
Mm.338933
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
161914
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pcdh15
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142735.1
NP_001136207.1
4
651
NM_001142736.1
NP_001136208.1
4
651
NM_001142737.1
NP_001136209.1
4
651
NM_001142738.1
NP_001136210.1
4
651
NM_001142739.1
NP_001136211.1
3
585
NM_001142740.1
NP_001136212.1
4
651
NM_001142741.1
NP_001136213.1
3
585
NM_001142742.1
NP_001136214.1
5
666
NM_001142743.1
NP_001136215.1
4
651
NM_001142746.1
NP_001136218.1
5
666
NM_001142747.1
NP_001136219.1
5
666
NM_001142748.1
NP_001136220.1
4
651
NM_001142760.1
NP_001136232.1
4
651
NM_023115.3
NP_075604.2
5
666
GenBank mRNA
AF281899.1
-
5
650
AK139154.1
-
5
591
DQ354396.1
-
4
635
DQ354397.1
-
4
635
DQ354398.1
-
4
635
DQ354399.1
-
4
635
DQ354400.1
-
4
635
DQ354401.1
-
4
635
DQ354402.1
-
3
569
DQ354403.1
-
4
635
DQ354404.1
-
3
569
DQ354405.1
-
5
650
DQ354406.1
-
4
635
DQ354408.1
-
4
635
DQ354409.1
-
4
635
DQ354410.1
-
4
635
DQ354411.1
-
4
635
DQ354412.1
-
4
635
DQ354413.1
-
5
650
DQ354414.1
-
4
635
DQ354416.1
-
5
591
DQ354417.1
-
4
576
DQ354418.1
-
4
576
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271377, XM_008772936.
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
startle response
morphogenesis of an epithelium
actin filament organization
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
visual perception
sensory perception of sound
locomotory behavior
adult walking behavior
adult locomotory behavior
nonmotile primary cilium assembly
multicellular organism growth
auditory receptor cell differentiation
photoreceptor cell maintenance
inner ear development
detection of mechanical stimulus involved in sensory perception of sound
sensory perception of light stimulus
equilibrioception
detection of mechanical stimulus involved in equilibrioception
response to calcium ion
righting reflex
auditory receptor cell stereocilium organization
inner ear receptor stereocilium organization
calcium ion binding
protein binding
protein complex binding
protein N-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline