Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142735.1 | NP_001136207.1 | 34 | 6737 | ||
NM_001142736.1 | NP_001136208.1 | 33 | 6731 | |||
NM_001142737.1 | NP_001136209.1 | 32 | 6524 | |||
NM_001142738.1 | NP_001136210.1 | 31 | 6611 | |||
NM_001142739.1 | NP_001136211.1 | 33 | 6671 | |||
NM_001142740.1 | NP_001136212.1 | 32 | 6722 | |||
NM_001142741.1 | NP_001136213.1 | 32 | 6662 | |||
NM_023115.3 | NP_075604.2 | 35 | 6752 | |||
GenBank mRNA | AF281899.1 | - | 35 | 6736 | ||
AK034124.1 | - | 1 | 1542 | |||
AK141024.1 | - | 1 | 1046 | |||
AY949849.1 | - | 22 | 4996 | |||
DQ354396.1 | - | 34 | 6721 | |||
DQ354397.1 | - | 33 | 6641 | |||
DQ354398.1 | - | 33 | 6715 | |||
DQ354399.1 | - | 32 | 6635 | |||
DQ354400.1 | - | 32 | 6508 | |||
DQ354401.1 | - | 31 | 6595 | |||
DQ354402.1 | - | 33 | 6655 | |||
DQ354403.1 | - | 32 | 6706 | |||
DQ354404.1 | - | 32 | 6646 |