Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252533.1 | NP_001239462.1 | 4 | 509 | ||
NM_023116.4 | NP_075605.1 | 5 | 640 | |||
GenBank mRNA | AK020806.1 | - | 4 | 585 | ||
AK034054.1 | - | 5 | 641 | |||
AK044560.1 | - | 4 | 1159 | |||
AK046048.1 | - | 4 | 497 | |||
AK078220.1 | - | 4 | 365 | |||
AK082315.1 | - | 5 | 524 | |||
AK143332.1 | - | 3 | 316 | |||
AM259383.2 | - | 4 | 509 | |||
BC109156.1 | - | 5 | 572 | 40047524 | ||
BC109157.1 | - | 5 | 572 | 40047531 | ||
FM872406.1 | - | 4 | 465 | |||
FM872407.1 | - | 4 | 571 | |||
FM872408.1 | - | 5 | 639 | |||
KJ789960.1 | - | 4 | 342 | |||
L20343.1 | - | 4 | 419 |