Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clns1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023671.2 | NP_076160.1 | 5 | 686 | ||
GenBank mRNA | AK011691.1 | - | 5 | 667 | ||
AK011789.1 | - | 5 | 672 | |||
AK031877.1 | - | 5 | 668 | |||
AK078050.1 | - | 5 | 639 | |||
AK133981.1 | - | 5 | 679 | |||
AK151356.1 | - | 5 | 671 | |||
AK151944.1 | - | 5 | 671 | |||
AK169259.1 | - | 5 | 650 | |||
AK205730.1 | - | 1 | 52 | |||
BC005575.1 | - | 5 | 652 | 3967933 | ||
U53455.1 | - | 5 | 598 |